AMBH2D700512 Ambeed, Inc.

(4-([1,1'-Biphenyl]-4-yl(phenyl)amino)phenyl)boronic acid

  • Cas Number : 1084334-86-0
  • Molecular Weight : 365.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1084334-86-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1084334-86-0.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart