AMBH2D7005BE Ambeed, Inc.

5-Chloro-1,3,3-trimethylspiro[indoline-2,3'-naphtho[2,1-b][1,4]oxazine]

  • MDL number MFCD00013319
  • Cas Number : 27333-50-2
  • Molecular Weight : 362.85

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/27333-50-2.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/27333-50-2.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart