AMBH2D7005D1 Ambeed, Inc.

Methyl 4-amino-2-isopropoxybenzoate

  • MDL number MFCD24624427
  • Cas Number : 909563-22-0
  • Molecular Weight : 209.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Inert atmosphere,Room temperature
Country of Origin China
Product Link https://www.ambeed.com/products/909563-22-0.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/909563-22-0.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart