AMBH2D7005D3 Ambeed, Inc.

2,2'-(Oxybis(4,1-phenylene))bis(1-phenylethane-1,2-dione)

  • MDL number MFCD00129168
  • Cas Number : 21454-19-3
  • Molecular Weight : 434.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 106
Purity 97
Flash Point 269.6
Storage temp. Sealed in dry,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 625.7�0�t 760 mmHg
Product Link https://www.ambeed.com/products/21454-19-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/21454-19-3.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart