AMBH2D7006F9 Ambeed, Inc.

1,3-Dimethoxypropan-2-amine

  • MDL number MFCD04038441
  • Cas Number : 78531-29-0
  • Molecular Weight : 119.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Flash Point 52
Storage temp. Keep in dark place,Inert atmosphere,2-8
Physical Form liquid
Shipping Temperature Normal
Country of Origin CN
Boiling point 160.8�t 760 mmHg
Product Link https://www.ambeed.com/products/78531-29-0.html

Safety Information

signalword Danger
pcodes P201-P261-P280-P310-P305+P351+P338
hcodes 8
MSDS https://www.ambeed.com/products/78531-29-0.html
pictograms GHS02,GHS05
hstatements H226-H314-H318
Availability details for
Quantity
  • Availability
  • Add to Cart