AMBH2D700709 Ambeed, Inc.

2-Amino-3-(methylamino)pyrazine

  • MDL number MFCD08460070
  • Cas Number : 84996-40-7
  • Molecular Weight : 124.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/84996-40-7.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/84996-40-7.html
pictograms GHS07
hstatements H302-H317-H319
Availability details for
Quantity
  • Availability
  • Add to Cart