AMBH2D7007EB Ambeed, Inc.

2-Amino-5-bromo-3-(diethylamino)pyrazine

  • MDL number MFCD08460064
  • Cas Number : 912773-09-2
  • Molecular Weight : 245.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/912773-09-2.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338-P310
MSDS https://www.ambeed.com/products/912773-09-2.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart