AMBH2D70080D Ambeed, Inc.

6-(Benzyloxy)-7H-purin-2-amine

  • MDL number MFCD00269931
  • Cas Number : 19916-73-5
  • Molecular Weight : 241.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Flash Point 329.6
Storage temp. Keep in dark place,Inert atmosphere,Store in freezer, under -20
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/19916-73-5.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/19916-73-5.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart