AMBH2D7008E3 Ambeed, Inc.

Quinoxalin-2-amine

  • MDL number MFCD00094018
  • Cas Number : 5424-05-5
  • Molecular Weight : 145.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 155-157
Purity 97
Flash Point 159
Storage temp. Keep in dark place,Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 297.6�t 760 mmHg
Product Link https://www.ambeed.com/products/5424-05-5.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/5424-05-5.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart