AMBH2D7008E9 Ambeed, Inc.

6'-(Diethylamino)-3'-methyl-2'-(phenylamino)-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one

  • MDL number MFCD00603523
  • Cas Number : 29512-49-0
  • Molecular Weight : 476.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 195-197
Purity 98
Flash Point 361.3
Storage temp. Keep in dark place,Sealed in dry,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 673.9�0�t 760 mmHg
Product Link https://www.ambeed.com/products/29512-49-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/29512-49-0.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart