AMBH2D700991 Ambeed, Inc.

5-Amino-2-bromopyrimidine

  • MDL number MFCD07778608
  • Cas Number : 56621-91-1
  • Molecular Weight : 174.00

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/56621-91-1.html

Safety Information

signalword Warning
pcodes P301+P312+P330
MSDS https://www.ambeed.com/products/56621-91-1.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart