AMBH2D7009AE Ambeed, Inc.

(R)-2-(2-Amino-3-(tritylthio)propanamido)acetic acid

  • Cas Number : 26988-61-4
  • Molecular Weight : 420.53

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 120
Purity 95
Flash Point 346.9
Storage temp. Keep in dark place,Inert atmosphere,Room temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 650.1�t 760 mmHg
Product Link https://www.ambeed.com/products/26988-61-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/26988-61-4.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart