AMBH2D7009CD Ambeed, Inc.

1-Ethyl-3,4-dihydroisoquinoline

  • MDL number MFCD02179236
  • Cas Number : 41173-70-0
  • Molecular Weight : 159.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 210
Purity 96
Storage temp. Sealed in dry,Room Temperature
Physical Form liquid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/41173-70-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/41173-70-0.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart