AMBH2D7009DB Ambeed, Inc.

2-Chloro-4-(pyridin-3-yl)pyrimidine

  • MDL number MFCD09743494
  • Cas Number : 483324-01-2
  • Molecular Weight : 191.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Flash Point 227.7
Storage temp. Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 399.5�t 760 mmHg
Product Link https://www.ambeed.com/products/483324-01-2.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/483324-01-2.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart