AMBH2D700E06 Ambeed, Inc.

((3AR,4R,6R,6aR)-6-(6-amino-2-chloro-9H-purin-9-yl)-2,2-dimethyltetrahydrofuro[3,4-d][1,3]dioxol-4-yl)methanol

  • MDL number MFCD09750845
  • Cas Number : 24639-06-3
  • Molecular Weight : 341.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 186-187
Synonym N/A
Purity 97
Flash Point 266.9
Storage temp. Keep in dark place,Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 517.7�t 760 mmHg
Product Link https://www.ambeed.com/products/24639-06-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/24639-06-3.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart