AMBH2D700E12 Ambeed, Inc.

2-Chloroisophthalonitrile

  • MDL number MFCD01657061
  • Cas Number : 28442-78-6
  • Molecular Weight : 162.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/28442-78-6.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/28442-78-6.html
pictograms GHS07
hstatements H302+H312+H332-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart