AMBH2D700E30 Ambeed, Inc.

2-Chloroquinoline-6-carbaldehyde

  • MDL number MFCD08236769
  • Cas Number : 791626-59-0
  • Molecular Weight : 191.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 165-167
Purity 97
Flash Point 161.5
Storage temp. Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 343.4�0�t 760 mmHg
Product Link https://www.ambeed.com/products/791626-59-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/791626-59-0.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart