AMBH2D700E96 Ambeed, Inc.

3,3',5,5'-Tetramethyl-[1,1'-biphenyl]-4,4'-diamine hydrochloride hydrate(1:2:x)

  • MDL number MFCD00150104
  • Cas Number : 207738-08-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Inert atmosphere,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/207738-08-7.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/207738-08-7.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart