AMBH2D700E99 Ambeed, Inc.

1-(2-Fluoro-4-methylphenyl)ethanone

  • MDL number MFCD03094331
  • Cas Number : 29427-48-3
  • Molecular Weight : 152.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/29427-48-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/29427-48-3.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart