AMBH2D700EBF Ambeed, Inc.

1-(2-Hydroxyethyl)-2,3,3-trimethyl-3H-indol-1-ium bromide

  • MDL number MFCD00980427
  • Cas Number : 29636-94-0
  • Molecular Weight : 284.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym N/A
Purity 97
Storage temp. Inert atmosphere,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/29636-94-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/29636-94-0.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart