AMBH2D700F03 Ambeed, Inc.

3-(Aminomethyl)pyridin-2(1H)-one hydrochloride

  • MDL number MFCD12035456
  • Cas Number : 85468-38-8
  • Molecular Weight : 160.60

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/85468-38-8.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/85468-38-8.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart