AMBH2D700F27 Ambeed, Inc.

(S)-7-Chloro-2,3-dihydrobenzofuran-3-amine hydrochloride

  • MDL number MFCD24435423
  • Cas Number : 2055848-78-5
  • Molecular Weight : 206.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/2055848-78-5.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/2055848-78-5.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart