AMBH2D700F5C Ambeed, Inc.

2-Isopropyl-4-nitrophenol

  • MDL number MFCD09749960
  • Cas Number : 60515-72-2
  • Molecular Weight : 181.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-isopropyl-4-nitrophenol
Inchi Code 1S/C9H11NO3/c1-6(2)8-5-7(10(12)13)3-4-9(8)11/h3-6,11H,1-2H3
Inchi Key YVSKFUMSIJTGTO-UHFFFAOYSA-N

Description

Synonym N/A
Purity 95%
Storage temp. Inert atmosphere,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/60515-72-2.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/60515-72-2.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart