AMBH2D700F8C Ambeed, Inc.

2-Cyclopropyl-5,6,7,8-tetrahydroimidazo[1,2-a]pyrazine hydrochloride

  • MDL number MFCD30730082
  • Cas Number : 2089380-98-1
  • Molecular Weight : 199.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym N/A
Purity 95
Storage temp. Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/2089380-98-1.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/2089380-98-1.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart