AMBH2D700F94 Ambeed, Inc.

Methyl 5-amino-5,6,7,8-tetrahydronaphthalene-2-carboxylate hydrochloride

  • MDL number MFCD22571285
  • Cas Number : 1097196-62-7
  • Molecular Weight : 241.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/1097196-62-7.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1097196-62-7.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart