AMBH2D700FBA Ambeed, Inc.

YL0919

  • MDL number MFCD30489758
  • Cas Number : 1339058-04-6
  • Molecular Weight : 334.84

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-((1-Benzyl-4-hydroxypiperidin-4-yl)methyl)pyridin-2(1H)-one hydrochloride
Purity 98
Storage temp. Inert atmosphere,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Product Link https://www.ambeed.com/products/1339058-04-6.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1339058-04-6.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart