AMBH2D700FC0 Ambeed, Inc.

LY303511 Hydrochloride

  • MDL number MFCD28167798
  • Cas Number : 2070014-90-1
  • Molecular Weight : 342.82

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/2070014-90-1.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/2070014-90-1.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart