AMBH2D700FC7 Ambeed, Inc.

CHIR-99021 Monohydrochloride

  • MDL number MFCD23704170
  • Cas Number : 1797989-42-4
  • Molecular Weight : 501.80

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1797989-42-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1797989-42-4.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart