AMBH2D700FEB Ambeed, Inc.

18-Methoxy-18-oxooctadecanoic acid

  • MDL number MFCD00673469
  • Cas Number : 72849-35-5
  • Molecular Weight : 328.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/72849-35-5.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/72849-35-5.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart