AMBH2D701055 Ambeed, Inc.

5(6)-Carboxy-Hexachlorofluorescein

  • MDL number MFCD31629689
  • Cas Number : 1166843-68-0
  • Molecular Weight : 582.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1166843-68-0.html

Safety Information

signalword Warning
pcodes P273
MSDS https://www.ambeed.com/products/1166843-68-0.html
pictograms GHS07
hstatements H302-H412
Availability details for
Quantity
  • Availability
  • Add to Cart