AMBH2D701061 Ambeed, Inc.

2-(2-(Chloromethyl)phenyl)acetic acid

  • MDL number MFCD06200800
  • Cas Number : 95335-46-9
  • Molecular Weight : 184.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 323.754�t 760 mmHg
Iupac Name [2-(chloromethyl)phenyl]acetic acid
Inchi Code 1S/C9H9ClO2/c10-6-8-4-2-1-3-7(8)5-9(11)12/h1-4H,5-6H2,(H,11,12)
Inchi Key RFYCQJHCAPCSTA-UHFFFAOYSA-N

Description

Purity 95%
Flash Point 149.601
Storage temp. Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 323.754�t 760 mmHg
Product Link https://www.ambeed.com/products/95335-46-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/95335-46-9.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart