AMBH2D70112A Ambeed, Inc.

4-Fluorothiophene-2-carboxylic acid

  • MDL number MFCD10566738
  • Cas Number : 32431-72-4
  • Molecular Weight : 146.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/32431-72-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/32431-72-4.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart