AMBH2D701153 Ambeed, Inc.

(2R,3R,4S,5R)-2-(2,6-Dichloro-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol

  • MDL number MFCD03411342
  • Cas Number : 13276-52-3
  • Molecular Weight : 321.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point >190°C
Synonym 2,6-Dichloropurineriboside
Purity 97
Storage temp. Inert atmosphere,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 589.3°C at 760 mmHg
Product Link https://www.ambeed.com/products/13276-52-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/13276-52-3.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart