AMBH2D7018A3 Ambeed, Inc.

3-(tert-Butoxy)-6-chloropyridazine

  • MDL number MFCD03425769
  • Cas Number : 17321-24-3
  • Molecular Weight : 186.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 97-99
Purity 97
Flash Point 135.4
Storage temp. Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 300.3�t 760 mmHg
Product Link https://www.ambeed.com/products/17321-24-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/17321-24-3.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart