AMBH2D7018FA Ambeed, Inc.

3,3-Dimethylmorpholine

  • MDL number MFCD09608072
  • Cas Number : 59229-63-9
  • Molecular Weight : 115.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Inert atmosphere,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/59229-63-9.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/59229-63-9.html
pictograms GHS07
hstatements H317-H319
Availability details for
Quantity
  • Availability
  • Add to Cart