AMBH2D701923 Ambeed, Inc.

3,4'-Dichlorodiphenyl ether

  • MDL number MFCD00043876
  • Cas Number : 6842-62-2
  • Molecular Weight : 239.10

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/6842-62-2.html

Safety Information

signalword Warning
pcodes P305+P351+P338
MSDS https://www.ambeed.com/products/6842-62-2.html
pictograms GHS07
hstatements H319
Availability details for
Quantity
  • Availability
  • Add to Cart