AMBH2D701927 Ambeed, Inc.

3,4-Dichlorostyrene

  • MDL number MFCD00013625
  • Cas Number : 2039-83-0
  • Molecular Weight : 173.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/2039-83-0.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/2039-83-0.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart