AMBH2D70195B Ambeed, Inc.

5-(2-Hydroxyethyl)-3,4-dimethylthiazol-3-ium iodide

  • MDL number MFCD00011958
  • Cas Number : 16311-69-6
  • Molecular Weight : 285.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Inert atmosphere,Room temperature
Country of Origin China
Product Link https://www.ambeed.com/products/16311-69-6.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/16311-69-6.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart