AMBH2D7019D4 Ambeed, Inc.

3,6-Dibromophenanthrene-9,10-dione

  • MDL number MFCD00451676
  • Cas Number : 53348-05-3
  • Molecular Weight : 366.00

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/53348-05-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/53348-05-3.html
pictograms GHS07
hstatements H302-H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart