AMBH2D7019EC Ambeed, Inc.

3-((tert-Butoxycarbonyl)amino)-3-(4-fluorophenyl)propanoic acid

  • MDL number MFCD02090715
  • Cas Number : 284493-72-7
  • Molecular Weight : 283.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/284493-72-7.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/284493-72-7.html
pictograms GHS07
hstatements H302-H312-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart