AMBH2D701A52 Ambeed, Inc.

Methyl 3-amino-4,5,6,7-tetrahydrobenzo[b]thiophene-2-carboxylate

  • MDL number MFCD05665152
  • Cas Number : 119205-38-8
  • Molecular Weight : 211.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Sealed in dry,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/119205-38-8.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/119205-38-8.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart