AMBH2D701A85 Ambeed, Inc.

5-Nitronicotinic acid

  • MDL number MFCD04038690
  • Cas Number : 2047-49-6
  • Molecular Weight : 168.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/2047-49-6.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/2047-49-6.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart