AMBH2D701B5F Ambeed, Inc.

3-Bromo-5-nitrobenzamide

  • MDL number MFCD01529620
  • Cas Number : 54321-80-1
  • Molecular Weight : 245.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/54321-80-1.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/54321-80-1.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart