AMBH2D701B82 Ambeed, Inc.

3-Bromophenylglyoxal hydrate

  • MDL number MFCD08705873
  • Cas Number : 106134-16-1
  • Molecular Weight : 231.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/106134-16-1.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/106134-16-1.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart