AMBH2D701B86 Ambeed, Inc.

3-Bromopropan-1-amine hydrobromide

  • MDL number MFCD00012912
  • Cas Number : 5003-71-4
  • Molecular Weight : 218.92

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/5003-71-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/5003-71-4.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart