AMBH2D701CD2 Ambeed, Inc.

3-Hydroxyadamantane-1-carboxylic acid

  • MDL number MFCD00193929
  • Cas Number : 42711-75-1
  • Molecular Weight : 196.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 203
Purity 97
Storage temp. Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 357.2�t 760 mmHg
Product Link https://www.ambeed.com/products/42711-75-1.html

Safety Information

signalword Warning
pcodes P305+P351+P338
MSDS https://www.ambeed.com/products/42711-75-1.html
pictograms GHS07
hstatements H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart