AMBH2D701D36 Ambeed, Inc.

3-(Boc-Amino)-3-(4-fluorophenyl)-1-propanol

  • MDL number MFCD04115551
  • Cas Number : 862466-16-8
  • Molecular Weight : 269.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 407.8�t 760 mmHg
Product Link https://www.ambeed.com/products/862466-16-8.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/862466-16-8.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart