AMBH2D702570 Ambeed, Inc.

4-(1H-imdazol-4-yl)piperidine

  • MDL number MFCD02179572
  • Cas Number : 106243-23-6
  • Molecular Weight : 151.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-(1H-Imdazol-4-yl)piperidine
Purity 98
Storage temp. Sealed in dry,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 368.1�t 760 mmHg
Product Link https://www.ambeed.com/products/106243-23-6.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/106243-23-6.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart