AMBH2D702594 Ambeed, Inc.

4-(2-Thiazolyl)phenol

  • MDL number MFCD07368615
  • Cas Number : 81015-49-8
  • Molecular Weight : 177.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/81015-49-8.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/81015-49-8.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart