AMBH2D7025DE Ambeed, Inc.

4-(Chlorodifluoromethoxy)aniline

  • MDL number MFCD01820809
  • Cas Number : 39065-95-7
  • Molecular Weight : 193.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Flash Point 94
Storage temp. Keep in dark place,Sealed in dry,2-8
Physical Form liquid
Shipping Temperature Normal
Country of Origin CN
Boiling point 231.9�t 760 mmHg
Product Link https://www.ambeed.com/products/39065-95-7.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/39065-95-7.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart